Order Entry
United States
ContactUsLinkComponent
RNaseAlert® Nuclease Control Reagents, Integrated DNA Technologies
RNaseAlert® Nuclease Control Reagents, Integrated DNA Technologies
Catalog # MSPP-11020102
CAS Number:  
RNaseAlert® Nuclease Control Reagents, Integrated DNA Technologies
Catalog # MSPP-11020102
Supplier Number:  11-02-01-02
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    RNaseAlert®-1 Kit
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-11020102

Specifications

About this item

These reagents are fluorescence-quenched oligonucleotide probes that emit a fluorescence signal only after nuclease degradation.

  • Used for qualitative detection of nuclease contamination
  • Used as quantitative tool to study enzyme kinetics
  • Employs a FAM™ reporter

The assay can be read visually or measured and quantified using fluorometry.

Delivery: It includes single use tubes, buffer, water, enzyme, and nuclease decontamination solution.