Left/Right Ergonomical Earmuffs, MSA
Headbands breakthrough design simulates the head's curves to provide a comfortable and stable fit, balancing earmuffs with the ear cup angle.
- Dedicated right and left cups are designed to fit ears and are shaped differently from front to back
- Ear cups can be adjusted individually, allowing close coverage even when they are not symmetrically placed on the head
Overall dimensions: 6.99" L x 4.33" W
: ANSI S3.19-1974.
Frequently Bought Together