Carry-Air with CO Monitor Systems, Allegro®
Supplier: Safety & Industrial Supplies
Certificates
About this item
A carbon monoxide monitor, which can be operated in AC or DC mode, activates an audible alarm when the concentration of CO exceeds 10 ppm (set for 5 ppm in Canada)
- 150 psig maximum inlet pressure
The Allegro® Air Filtration Panels convert shop air used for pneumatic tools and equipment to clean breathable air. The 3-stage filtration system purifies compressed air removing water, oil, and particulate contaminants.
Frequently Bought Together