Riffle Pan, New Liskeard Sheet Metal
Catalog # 11019-604
Supplier: NEW LISKEARD SHEET METAL WORKS
Specifications
- Description:Riffle Pan
- Cat. No.:11019-604
- Supplier no.:SP111
Specifications
About this item
Pan is designed to facilitate rapid and even splitting of solid samples and other dry substances.
Pan dimensions are 12 x 5 x 3.5".