R-Phycoerythrin, Pierce™
Catalog # PI46185
Supplier: Avantor
CAS Number:
Specifications
- Size:2 mg
- Cat. No.:PI46185
- Supplier no.:46185
Specifications
About this item
:
:240 kDa
Storage Temperature:Refrigerator
CAS Number:11016-17-4
MDL Number:MFCD00081950