Molecular Model Set, 31 Atoms
Catalog # 470231-428
Supplier: Wards
Specifications
- Description:Molecular Model Set, 31 Atoms
- Cat. No.:470231-428
Specifications
About this item
Suitable for VSEPR Study
- 31 atoms
- 20 short links
This molecular model set allows students to build multiple models of simple molecules in their compact representation. It is possible to present molecules acting as reagents or products of simple chemical reactions and to achieve a molecular representation of pure or polluted air. It is also suitable for a first study of the geometry of molecules (VSEPR).
Contents: 12 White 1 Face Hydrogen (30mm), 4 Black 4 Face Carbon (38 mm), 1 Black 2 Face Carbon (38 mm), 2 Red 2 Face Oxygen (38 mm), 7 Red 1 Face Oxygen (38 mm), 1 Blue 3 Face Nitrogen (38 mm), 2 Blue 1 Face Nitrogen (38 mm), 2 Green 1 Face Chlorine (50 mm), 20 Short Links (16 mm).