Orders LinkContactUsLinkComponent
Proto® Phillips Socket Bits, ORS Nasco
Proto® Phillips Socket Bits, ORS Nasco
Certificates
Proto® Phillips Socket Bits, ORS Nasco
Proto® Phillips Socket Bits, ORS Nasco
5 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Material
    Length
    Color
    Drive size
    Availability
    Price
  • 300088-688
    Ball Locking, #2 Phillips® Tip
    Ball Locking, #2 Phillips® Tip
    Forged alloy steel
    1.9687"
    Nickel chrome
    0.375" Square
    Log in for availability
    Log in for availability
    Each
    Each - $26.86
    $26.86
    Each
       $26.86
    Supplier Number: 577-5241
  • 300088-690
    Ball Locking, #3 Phillips® Tip
    Ball Locking, #3 Phillips® Tip
    Forged alloy steel
    1.9687"
    Nickel chrome
    0.375" Square
    Log in for availability
    Log in for availability
    Each
    Each - $28.63
    $28.63
    Each
       $28.63
    Supplier Number: 577-5242
  • 300025-488
    Ball Locking, #2 Phillips® Tip
    Ball Locking, #2 Phillips® Tip
    Forged alloy steel
    1.0625"
    Nickel chrome
    0.25" Square
    Log in for availability
    Log in for availability
    Each
    Each - $23.80
    $23.80
    Each
       $23.80
    Supplier Number: 577-4738
  • 300025-486
    Ball Locking, #1 Phillips® Tip
    Ball Locking, #1 Phillips® Tip
    Forged alloy steel
    1.0625"
    Nickel chrome
    0.25" Square
    Log in for availability
    Log in for availability
    Each
    Each - $23.11
    $23.11
    Each
       $23.11
    Supplier Number: 577-4737
  • 300088-692
    Ball Locking, #4 Phillips® Tip
    Ball Locking, #4 Phillips® Tip
    Forged alloy steel
    1.9687"
    Nickel chrome
    0.375" Square
    Log in for availability
    Log in for availability
    Each
    Each - $26.86
    $26.86
    Each
       $26.86
    Supplier Number: 577-5243