The fastest method for complete bisulfite conversion of DNA for methylation analysis.
- Fastest method for complete bisulfite conversion of DNA for methylation analysis
- Ready-to-use conversion reagent is added directly to DNA
- High-yield, converted DNA is ideal for PCR, MSP, array, bisulfite and Next-Generation sequencing
- >99.5% conversion efficiency
Bisulfite conversion is considered the gold standard in DNA methylation analysis. The only downside is that the bisulfite conversion process is relatively harsh and will innately damage the DNA, leading to DNA fragmentation and low recovery. The EZ DNA Methylation – Lightning® Kit features the fastest bisulfite conversion method resulting in fully converted DNA with reduced fragmentation and more efficient PCR amplification. The bisulfite converted DNA is ideal for downstream DNA methylation analyses such as PCR, MSP, array, bisulfite and Next-Generation sequencing.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: