Pierce™ Avidin, FITC (Fluorescein Isothiocyanate)
Catalog # PI21221
Supplier: Invitrogen
CAS Number:
Specifications
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Size:5 mg
- Storage Conditions:Store at 2 – 8 °C
- Protein/Peptide Name:Avidin
- Cat. No.:PI21221
- Supplier no.:21221
Specifications
About this item
Pierce™ FITC conjugated avidin, is a purified avidin protein from hen egg whites that effectively binds biotin, a tag that is frequently used to tag antibodies and other probes for immunodetection methods.
- Native avidin - glycoprotein from chicken egg whites, 67 kDa, pI = 10
- Glycosylation and isoelectric point renders avidin more soluble than streptavidin
- Significantly less expensive than recombinant streptavidin
- Avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
Application: Fluorescence-Activated Cell Sorting and Histochemical Staining