Pierce SuperSignal™ ELISA Pico Chemiluminescent Substrate, Thermo Scientific
SuperSignal™ ELISA Pico chemiluminescent substrate is optimized to generate an intense light signal and provide exceptional performance in luminometer based assays.
- Intense signal is produced immediately at room temperature or at 37 °C Signal can be read in black or white opaque microplates Detects protein in ELISAs down to the picogram levels Ambient shipping and room temperature storage Consistent performance of the working solution over eight hour period
It offers greater sensitivity in ELISAs, or any other solution-based assay using chemiluminescent substrates for enzyme detection and quantification. These ELISAs can take place in either a test tube or a microplate and are quantified by measuring relative light units (RLU) in a luminometer.
- Sensitivity:Picogram (10-12)
Frequently Bought Together