Order Entry
United States
ContactUsLinkComponent
 
Quick-DNA™ Plant/Seed Kits, Zymo Research
 
undefined
Quick-DNA™ Plant/Seed Kits, Zymo Research

 

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    Quick-DNA™ Plant/Seed 96 Kit

 

Frequently Bought Together