


n-Nonane 99%, Reagent Grade
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:111-84-2
:
Formula:H₃C(CH₂)₇CH₃
MDL Number:MFCD00009574
Boiling Pt:2.9 %(V)
Melting Pt:–53 °C
Density:0.72 g/cm³ (20 °C)
Flash Pt:31 °C
Storage Temperature:Ambient
EINECS:203-913-4
ADR:3,III
:128.26 g/mol
UN:1920
:GHS07
:GHS08
:GHS02
Specification Test Results
- Explosion Limit:2.9 %
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Refractive index (n 20/D):1.403 - 1.407
- Autoignition Temperature:401 °F
- Vapor Density:4.41 (vs air)
Specification Test Results
Frequently Bought Together