Order Entry
United States
ContactUsLinkComponent
2-Propanol ≥99.0%, meets analytical specification of USP, Riedel-de Haën™
2-Propanol ≥99.0%, meets analytical specification of USP, Riedel-de Haën™
Catalog # BJI9030-500ML
CAS Number:  
undefined
2-Propanol ≥99.0%, meets analytical specification of USP, Riedel-de Haën™
Catalog # BJI9030-500ML
Supplier Number:  I9030-500ML
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Size:
    500 ml
  • Environmentally Preferable:
  • Cat. No.:
    BJI9030-500ML

Specifications

Specification Test Results

  • Vapor Density:
    2.1 (vs air)
  • Assay (GC):
    Min. 99.0 %
  • Density (D 25/25):
    0.783 - 0.787
  • Refractive index (n 20/D):
    1.376 - 1.378
  • Water (Karl Fischer):
    Max. 0.5 %
  • Non-volatile matter:
    Max. 0.005 %
  • Acidity:
    complying
  • Volatile impurities (GC):
    complying
  • Acetone (GC):
    Max. 0.1 %
  • 2-Butanol (GC):
    Max. 0.1 %
  • Diethyl ether (GC):
    Max. 0.1 %
  • Diisopropyl alcohol:
    Max. 0.1 %
  • 1-propanol (GC):
    Max. 0.1 %
  • Impurities (GC) total:
    Max. 1.0 %
  • Residual solvents:
    complying

Specification Test Results

About this item

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
CAS Number:67-63-0
 :
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
 :60.1 g/mol
Boiling Pt:77…83 °C (1.013 hPa)
Boiling Pt:82
Melting Pt:–90 °C
Melting Pt:-89
Density:0.784…0.786 g/cm³ (20 °C)
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :GHS07
 :GHS02
ADR:3 II
Merck Index:13,05228