Order Entry
United States
ContactUsLinkComponent
 
Flammable
Irritant
Acetone ≥99.9%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Catalog # BJ69508-1L
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Acetone ≥99.9%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Catalog # BJ69508-1L
Supplier Number:  69508-1L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    Chem21 solvent selection guide recommended after discussion
  • Cat. No.:
    BJ69508-1L

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Water (Karl Fischer):
    Max. 0.20 %
  • Silver (Ag):
    Max. 0.5 ppb
  • Aluminium (Al):
    Max. 1 ppb
  • Arsenic (As):
    Max. 1 ppb
  • Gold (Au):
    Max. 0.5 ppb
  • Barium (Ba):
    Max. 0.5 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • Bismuth (Bi):
    Max. 0.5 ppb
  • Calcium (Ca):
    Max. 20 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 0.5 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 1 ppb
  • Iron (Fe):
    Max. 5 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Germanium (Ge):
    Max. 0.5 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Iridium (Ir):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 1 ppb
  • Manganese (Mn):
    Max. 0.5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 20 ppb
  • Nickel (Ni):
    Max. 1 ppb
  • Lead (Pb):
    Max. 0.5 ppb
  • Palladium (Pd):
    Max. 0.5 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 0.5 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Tin (Sn):
    Max. 0.5 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 1 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 0.5 ppb
  • Zinc (Zn):
    Max. 5 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Bromide (Br):
    Max. 10 ppb
  • Chloride (Cl):
    Max. 25 ppb
  • Fluoride (F):
    Max. 10 ppb
  • Iodide (I):
    Max. 10 ppb
  • Nitrite (NO₂):
    Max. 10 ppb
  • Nitrate (NO₃):
    Max. 25 ppb
  • Phosphate (PO₄):
    Max. 10 ppb
  • Sulfate (SO₄):
    Max. 50 ppb
  • Aldehydes (as HCHO):
    Max. 10 ppm
  • Methanol (GC):
    Max. 0.05 %
  • 2-propanol (GC):
    Max. 0.05 %
  • Vapor Density:
    2 (vs air)
  • Refractive Index:
    n20/D 1.359 (lit.)

Specification Test Results

About this item

CAS Number:67-64-1
 :
Formula:CH₃COCH₃
MDL Number:MFCD00008765
 :58.08 g/mol
Boiling Pt:14.3 %(V)
Melting Pt:–95 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:–17 °C
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
 :GHS02
 :GHS07