

Methanol, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # BJ646377-4X4L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJ646377-4X4L
Specifications
Specification Test Results
- Purity (GC):Min. 99.90 %
- IR spectrum:complying
- Non-volatile matter:Max. 0.0001 %
- Water (Karl Fischer):Max. 0.05 %
- Titrable Acid:Max. 0.0003 meq/g
- Titrable Base:Max. 0.0002 meq/g
- Absorbance at 400 nm:Max. 0.005
- Absorbance at 250 nm:Max. 0.02
- Absorbance at 230 nm:Max. 0.10
- Absorbance at 220 nm:Max. 0.20
- Absorbance at 210 nm:Max. 0.80
- Absorbance at 205 nm:Max. 1.0
- Fluorescence (chinin) at 254 nm:Max. 1.0 ppb
- Fluorescence (chinin) at 365 nm:Max. 1.0 ppb
- HPLC Gradient at 230 nm:Max. 2 mAU
- HPLC Gradient at 254 nm:Max. 5 mAU
- APHA:Max. 10
- Carbonyl compounds (as Acetone):Max. 0.001 %
- Carbonyl Compounds (as HCHO):Max. 0.001 %
- Carbonyl compounds (as C₂H₄O):Max. 0.001 %
- Explosion Limit:36 %
- Autoignition Temperature:725 °F
Specification Test Results
About this item
:
:32.04 g/mol
Boiling Pt:64.7 °C
Melting Pt:-98 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:11 °C
UN:1230
ADR:3,II
:GHS08
:GHS02
Storage Temperature:Ambient
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595