Order Entry
United States
ContactUsLinkComponent
Iodine ≥99.5% (RT), particles (spherical), Puriss., Fluka™
Irritant
Health
Iodine ≥99.5% (RT), particles (spherical), Puriss., Fluka™
Catalog # BJ57655-1KG
CAS Number:  
Irritant
Health
Iodine ≥99.5% (RT), particles (spherical), Puriss., Fluka™
Catalog # BJ57655-1KG
Supplier Number:  57655-1KG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 kg
  • Cat. No.:
    BJ57655-1KG

Specifications

Specification Test Results

  • Assay:
    99.5 - 100.5 %
  • Appearance of the substance:
    complying
  • Vapor Pressure:
    0.31 mmHg ( 25 °C)
  • Form:
    particles (round)
  • Resistivity:
    1.3E15 μΩ-cm
  • Vapor Density:
    9 (vs air)

Specification Test Results

About this item

CAS Number:7553-56-2
 :
Formula:I₂
MDL Number:MFCD00011355
 :253.81 g/mol
Melting Pt:113.5 °C
Density:4.930 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8,III
 :GHS07
 :GHS08