


1-Butanol ≥99.9%
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Boiling Pt:119 °C (1.013 hPa)
Melting Pt:<–90 °C
Density:0.81 g/cm³ (20 °C)
Flash Pt:9.4 %(V)
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
:GHS02
:GHS05
:GHS07
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.80 %
- Density (D 20/4):0.809 - 0.811
- Non-volatile matter:Max. 20 mg/l
- Water (Karl Fischer):Max. 0.1 %
- Free acid (as CH₃COOH):Max. 0.005 %
- Iron (Fe):Max. 0.3 ppm
- Di-n-butyl ether (GC):Max. 0.100 %
- Isobutyl alcohol (GC):Max. 0.100 %
- n-Butyraldehyde (GC):Max. 0.020 %
- Carbonyl compounds (as CO):Max. 0.050 %
- APHA:Max. 10
- Autoignition Temperature:649 °F
- Refractive Index:n20/D 1.399 (lit.)
Specification Test Results
Related Information
Frequently Bought Together