


Xylene (mixture of isomers) for histology
Catalog # BJ534056-4L
Supplier: Honeywell Research Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Aluminum bottle
- Size:4 L
- Cat. No.:BJ534056-4L
Specifications
Specification Test Results
- Vapor Pressure:18 mmHg ( 37.7 °C)
- IR spectrum:complying
- C₈ isomers including Ethylbenzene (GC):Min. 96 %
- Total isomers (GC):Min. 73 %
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.05 %
- Ethyl benzene (GC):Max. 25.0 %
- APHA:Max. 10
Specification Test Results
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Boiling Pt:136 - 140 °C
Boiling Pt:136…140 °C
Melting Pt:-34 °C
Melting Pt:–34 °C
Density:0.87 g/cm³
Flash Pt:25 °C
UN:1307
ADR:3,III
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient