

Nitric acid concentrate for dilution to 1L, 1.0 M HNO3 (1.0N) volumetric solution, Fluka™
:7697-37-2
:
:HNO₃
:MFCD00011349
:63.01
:100 °C
:no data available
:1.200 g/cm³ (20 °C)
:Ambient
:2031
:8,II
:GHS06
:GHS05
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Frequently Bought Together