Order Entry
United States
ContactUsLinkComponent
Potassium permanganate concentrate for dilution to 1L, 1/50 M KMnO4 (0.1N) volumetric solution, Fluka™
Corrosive
Potassium permanganate concentrate for dilution to 1L, 1/50 M KMnO4 (0.1N) volumetric solution, Fluka™
Catalog # BJ38130-6X1EA
CAS Number:  
Corrosive
Potassium permanganate concentrate for dilution to 1L, 1/50 M KMnO4 (0.1N) volumetric solution, Fluka™
Catalog # BJ38130-6X1EA
Supplier Number:  38130-6X1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Size:
    6 x 1 Each
  • Cat. No.:
    BJ38130-6X1EA

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

CAS Number:7722-64-7
 :
Formula:KMnO₄
MDL Number:MFCD00011364
 :158.03 g/mol
 :GHS05
Storage Temperature:Ambient