


Potassium dichromate concentrate for dilution to 1L, 1/60 M K2Cr2O7 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7778-50-9
:
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367
:294.18 g/mol
Boiling Pt:100 °C (1.013 hPa)
Melting Pt:no data available
Density:1.030 g/cm³ (20 °C)
UN:2922
ADR:8,II
:GHS05
:GHS08
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
Frequently Bought Together