Acetonitrile with 0.1% (v/v) trifluoroacetic acid ≥99.5%, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:76-05-1
:75-05-8
MDL Number:MFCD00001878
Boiling Pt:81…82 °C
Melting Pt:no data available
Density:0.78 g/cm³ (20 °C)
Flash Pt:6 °C
EINECS:200-835-2
UN:1648
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended by default
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- Assay trifluoroacetic acid:0.093 - 0.107 Vol.%
- Calcium (Ca):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.5 ppm
- Sodium (Na):Max. 2 ppm
- Transmittance at 210 nm:Min. 30 %
- Transmittance at 230 nm:Min. 50 %
- Transmittance from 254 nm:Min. 90 %
- HPLC Gradient at 254 nm:Max. 50 mAU
- LC-MS application:complying
Specification Test Results
Related Information
Frequently Bought Together