ContactUsLinkComponent
Perchloric acid 0.01 M HClO4 (0.01N) in acetic acid volumetric solution, for titration in non-aqueous liquids, Fluka™
Perchloric acid 0.01 M HClO4 (0.01N) in acetic acid volumetric solution, for titration in non-aqueous liquids, Fluka™
  BJ34408-1L
 :  
undefined
Perchloric acid 0.01 M HClO4 (0.01N) in acetic acid volumetric solution, for titration in non-aqueous liquids, Fluka™
  BJ34408-1L
 :  34408-1L
 :  
  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ34408-1L
  • c(perchloric acid):
    9.95 - 10.05 mM
  • Water (Karl Fischer):
    0.05 - 0.15 %
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective
 :7601-90-3
 :
 :HClO₄
 :MFCD00011325
 :100.46 g/mol
 :119 °C (1.013 hPa) (solvent)
 :no data available
 :1.06 g/cm³ (20 °C)
 :40 °C (The information regarding the flash point is that of the solvent)
 :40 °C (solvent)
 :Ambient
 :2920
 :8,II
 :GHS05
 :GHS02