


Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:–46 °C
Density:0.7857 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
:GHS08
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended by default
Specifications
Specification Test Results
- Explosion Limit:16 %
- Appearance:complying
- Assay (GC):Min. 99.5 %
- Boiling range:81 - 82 °C
- Boiling range 80-82 °C:Min. 95 %
- Density (D 20/4):0.781 - 0.783
- Refractive index (n 20/D):1.3435 - 1.3445
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.1 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Titrable Acid:Max. 8 µeq/g
- Free alkali (as NH₃):Max. 0.001 %
- Titrable Base:Max. 0.6 µeq/g
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- APHA:Max. 10
Specification Test Results
Related Information
Frequently Bought Together