


Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # BJ293253-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ293253-1L
Specifications
Specification Test Results
- Autoignition Temperature:437 °F
- Identity (IR):complying
- Assay of hexane (all isomers, GC):Min. 98.50 %
- Refractive index (n 20/D):1.377 - 1.382
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.02 %
- Titrable Acid:Max. 0.0003 meq/g
- S-compounds (as S):Max. 0.005 %
- Thiophene:complying
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 250 nm:Max. 0.01
- Absorbance at 240 nm:Max. 0.02
- Absorbance at 230 nm:Max. 0.05
- Absorbance at 220 nm:Max. 0.10
- Absorbance at 210 nm:Max. 0.30
- Absorbance at 195 nm:Max. 1.0
- APHA:Max. 10
- Vapor Density:3 (vs air)
- Explosion Limit:7.5 %
Specification Test Results
About this item
CAS Number:110-54-3
:
Formula:H₃C(CH₂)₄CH₃
:86.18 g/mol
Boiling Pt:48…65 °C (1.013 hPa)
Melting Pt:–154 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–26 °C
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
:92112-69-1
MDL Number:MFCD02179311