


Alcohol, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Catalog # BJ270741-4L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4x 4 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA safer chemical solvent (Green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Cat. No.:BJ270741-4L
- Supplier no.:270741-4X4L
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 89.0 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.1 %
- Methanol (GC):3.0 - 5.0 %
- 2-propanol (GC):4.0 - 6.0 %
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 300 nm:Max. 0.01
- Absorbance at 255 nm:Max. 0.04
- Absorbance at 220 nm:Max. 0.35
- Absorbance at 210 nm:Max. 0.65
- Absorbance at 205 nm:Max. 1.00
Specification Test Results
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
MDL Number:MFCD00003568
Boiling Pt:78.32 °C (main component)
Melting Pt:–114.1 °C (main component)
Density:0.7890 g/cm³ (20 °C)
Flash Pt:15 °C (main component)
EINECS:200-578-6
UN:1987
ADR:3,II
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient