


Carbon disulfide ≥99.9% (by GC) ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:75-15-0
:
Formula:CS₂
MDL Number:MFCD00011321
:76.14 g/mol
Boiling Pt:46 °C (1.013 hPa)
Melting Pt:–76 °C
Density:1.264 g/cm³ (20 °C)
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:200-843-6
UN:1131
ADR:3,I
:GHS02
:GHS08
:GHS07
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.05 %
- Sulfur dioxide (SO₂):complying
- Hydrogen sulfide (H₂S):complying
- APHA:Max. 10
- Autoignition Temperature:212 °F
- Refractive Index:n20/D 1.627 (lit.)
- Explosion Limit:50 %
Specification Test Results
Frequently Bought Together