Acetonitrile with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Catalog # BJ14272-2L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2 L
- Cat. No.:BJ14272-2L
Specifications
Specification Test Results
- Formic acid:0.095 - 0.105 Vol.%
- Non-volatile matter:Max. 0.0001 %
- Water (Karl Fischer):Max. 0.02 %
- Aluminium (Al):Max. 0.03 ppm
- Calcium (Ca):Max. 0.1 ppm
- Iron (Fe):Max. 0.05 ppm
- Potassium (K):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.03 ppm
- Sodium (Na):Max. 0.1 ppm
- Transmittance at 254 nm:Min. 90 %
- Transmittance at 230 nm:Min. 15 %
- Transmittance at 210 nm:Min. 5 %
- Suitability for the UHPLC-MS:complying
- UHPLC Gradient drift 254 nm:Max. 30 mAU
- UHPLC Gradient peak 254 nm:Max. 2 mAU
- UHPLC-MS ESI(+) (Reserpine):Max. 20 ppb
- Fluorescence (chinin) at 365 nm:Max. 0.5 ppb
- Fluorescence (chinin) at 254 nm:Max. 0.5 ppb
Specification Test Results
About this item
:75-05-8
Boiling Pt:81 °C
Melting Pt:–46 °C
Density:0.78 g/cm³ (20 °C)
Flash Pt:5 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
:GHS02
:GHS08
:GHS07