


1,2-Dichloroethane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:107-06-2
:
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963
:98.96 g/mol
Boiling Pt:83 °C
Melting Pt:–35 °C
Density:1.25 g/cm³ (25 °C)
Flash Pt:13 °C.1.013 hPa
Storage Temperature:Ambient
EINECS:203-458-1
UN:1184
ADR:3,II
:GHS08
:GHS06
:GHS02
Specification Test Results
- Assay (GC):Min. 99.5 %
- Identity (IR):complying
- Refractive index (n 20/D):1.443 - 1.445
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.03 %
- Free acid (as HCl):Max. 0.001 %
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.05 ppm
- Iron (Fe):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- APHA:Max. 25
- Vapor Pressure:87 mmHg ( 25 °C)
Specification Test Results
Frequently Bought Together