


Nitric acid 67-69% (by titrimetric analysis), TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01
Boiling Pt:122 °C (1.013 hPa)
Melting Pt:–38 °C
Density:1.420 g/cm³ (20 °C)
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS03
:GHS05
:GHS06
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay:64.5 - 70 %
- Silver (Ag):Max. 10 ppt
- Aluminium (Al):Max. 50 ppt
- Arsenic (As):Max. 100 ppt
- Gold (Au):Max. 20 ppt
- Barium (Ba):Max. 10 ppt
- Beryllium (Be):Max. 10 ppt
- Bismuth (Bi):Max. 10 ppt
- Calcium (Ca):Max. 100 ppt
- Cadmium (Cd):Max. 10 ppt
- Cerium (Ce):Max. 10 ppt
- Cobalt (Co):Max. 10 ppt
- Chromium (Cr):Max. 50 ppt
- Cesium (Cs):Max. 10 ppt
- Copper (Cu):Max. 20 ppt
- Dysprosium (Dy):Max. 10 ppt
- Erbium (Er):Max. 10 ppt
- Europium (Eu):Max. 10 ppt
- Iron (Fe):Max. 100 ppt
- Gallium (Ga):Max. 10 ppt
- Gadolinium (Gd):Max. 10 ppt
- Germanium (Ge):Max. 10 ppt
- Hafnium (Hf):Max. 10 ppt
- Mercury (Hg):Max. 500 ppt
- Holmium (Ho):Max. 10 ppt
- Indium (In):Max. 10 ppt
- Potassium (K):Max. 100 ppt
- Lanthanum (La):Max. 10 ppt
- Lithium (Li):Max. 10 ppt
- Lutetium (Lu):Max. 10 ppt
- Magnesium (Mg):Max. 50 ppt
- Manganese (Mn):Max. 10 ppt
- Molybdenum (Mo):Max. 10 ppt
- Sodium (Na):Max. 100 ppt
- Niobium (nb):Max. 10 ppt
- Neodymium (nd):Max. 10 ppt
- Nickel (Ni):Max. 50 ppt
- Lead (Pb):Max. 10 ppt
- Palladium (Pd):Max. 50 ppt
- Praseodymium (Pr):Max. 10 ppt
- Platinum (Pt):Max. 20 ppt
- Rubidium (Rb):Max. 10 ppt
- Rhenium (Re):Max. 10 ppt
- Rhodium (Rh):Max. 10 ppt
- Antimony (Sb):Max. 10 ppt
- Scandium (Sc):Max. 50 ppt
- Selenium (Se):Max. 100 ppt
- Samarium (Sm):Max. 10 ppt
- Tin (Sn):Max. 20 ppt
- Strontium (Sr):Max. 10 ppt
- Terbium (Tb):Max. 10 ppt
- Tellurium (Te):Max. 10 ppt
- Thorium (Th):Max. 10 ppt
- Tantalum (Ta):Max. 50 ppt
- Titanium (Ti):Max. 10 ppt
- Thallium (Tl):Max. 10 ppt
- Uran (U):Max. 10 ppt
- Vanadium (V):Max. 10 ppt
- Tungsten (W):Max. 10 ppt
- Yttrium (Y):Max. 10 ppt
- Ytterbium (Yb):Max. 10 ppt
- Zinc (Zn):Max. 50 ppt
- Zirconium (Zr):Max. 10 ppt
Specification Test Results
Related Information
Frequently Bought Together