Order Entry
United States
ContactUsLinkComponent
Bovine Rhodopsin Epitope Tag
Bovine Rhodopsin Epitope Tag
Catalog # 103007-234
Supplier:  Anaspec Inc
CAS Number:  
Bovine Rhodopsin Epitope Tag
Catalog # 103007-234
Supplier:  Anaspec Inc
Supplier Number:  AS-62190
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Source:
  • Species:
    Bovine
  • Size:
    5 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Rhodopsin tag
  • Protein/Peptide Name:
    Rhodopsin Epitope Tag
  • Purity:
    95%
  • Molecular Weight:
    903 Da
  • Sequence:
    TETSQVAPA
  • Cat. No.:
    103007-234
  • Supplier no.:
    AS-62190

Specifications

About this item

This is a 9-amino acid peptide corresponding to the C-terminus of bovine rhodopsin. It is widely used as an epitope tag. A number of anti-rhodopsin antibodies recognize this epitope.
Sequence:TETSQVAPA
MW:903 Da
% peak area by HPLC:95
Storage condition:-20° C