Order Entry
United States
Orders LinkContactUsLinkComponent
W Peptide

W Peptide

1 mg
Catalog # 102996-752
Marketsource
Supplier:   Anaspec
Supplier #:  AS-27069
W Peptide
Catalog # 102996-752
Supplier:  Anaspec
Marketsource
CAS Number:  
W Peptide

1 mg

CAS Number:  
Price
$130.08
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This peptide, containing the consensus sequence XKYX(P/V)M, is found to stimulate phospholipase C (PLC)-mediated formation of InoPs in certain cell lines and human neutrophils. WKYMVm-NH2 may have the ability to activate the microbicidal functions of human neutrophils.
Sequence:WKYMVm-NH2
MW:856.1 Da
% peak area by HPLC:95
Storage condition:-20° C

Documents

Specifications

Specifications

  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Cat. No.:
    102996-752
  • Molecular Weight:
    856.1 Da
  • Purity:
    95%
  • Protein Synonyms:
    Neutrophil activating Peptide
  • Protein/Peptide Name:
    W Peptide
  • Conjugation:
    Unconjugated
  • Sequence:
    WKYMVm-NH2

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.