Order Entry
United States
ContactUsLinkComponent
Suc-LLVY, AMC (7-amino-4-methylcoumarin)
Suc-LLVY, AMC (7-amino-4-methylcoumarin)
Catalog # 103007-798
Supplier:  Anaspec Inc
CAS Number:  
Suc-LLVY, AMC (7-amino-4-methylcoumarin)
Catalog # 103007-798
Supplier:  Anaspec Inc
Supplier Number:  AS-63892
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    AMC (7-amino-4-methylcoumarin)
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Calpain substrate|20S proteasome substrate
  • Protein/Peptide Name:
    Suc-LLVY-AMC
  • Purity:
    95%
  • Molecular Weight:
    763.9 Da
  • Sequence:
    Suc-LLVY-AMC
  • Cat. No.:
    103007-798
  • Supplier no.:
    AS-63892

Specifications

About this item

Calpains are a family of intracellular Ca2+ dependent cysteine proteases. They respond to Ca2+ signals by cleaving many specific proteins, thereby irreversibly modifying their function(s). They are implicated in a variety of Ca2+ regulated cellular processes as well as various pathological phenomena, such as ischemic injury, muscular dystrophy, diabetes, cataract, atherosclerosis, Alzheimer’s disease, and cancer. Calpains represent potential therapeutic targets for drug discovery.
This 7-amino-4-methylcoumarin (AMC) labeled peptide is widely used as a fluorogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases. Cleavage of this AMC peptide by the enzymes generates strongly fluorescent AMC that is monitored fluorimetrically at Abs/Em=353/442 nm.
Sequence:Suc-LLVY-AMC
MW:763.9 Da
% peak area by HPLC:95
Storage condition:-20° C