Order Entry
United States
ContactUsLinkComponent
Myristolated PKC Zeta
Myristolated PKC Zeta
Catalog # 103007-678
Supplier:  Anaspec
CAS Number:  
Myristolated PKC Zeta
Catalog # 103007-678
Supplier:  Anaspec
Supplier Number:  AS-63695
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Myristoylated PKCζ pseudosubstrate Peptide|PKC zeta Peptide|PKC-derived ζ-inhibitory Peptide (ZIP)|Protein kinase zeta inhibitor
  • Protein/Peptide Name:
    Myristolated PKC Zeta
  • Purity:
    95%
  • Molecular Weight:
    1928.5 Da
  • Sequence:
    Myr-RLYRKRIWRSAGR
  • Cat. No.:
    103007-678
  • Supplier no.:
    AS-63695

Specifications

About this item

This is a myristoylated PKCζ pseudosubstrate-derived ζ-inhibitory peptide (ZIP) found to inhibit an atypical type of Protein Kinase C ζ (zeta)
Sequence:Myr-RLYRKRIWRSAGR
MW:1928.5 Da
% peak area by HPLC:95
Storage condition:-20° C