Order Entry
United States
Orders LinkContactUsLinkComponent
CREBtide

CREBtide

5 mg
Catalog # 103003-190
Marketsource
Supplier:   Anaspec
Supplier #:  AS-60510-5
CREBtide
Catalog # 103003-190
Supplier:  Anaspec
Marketsource
CAS Number:  
CREBtide

5 mg

CAS Number:  
Price
$936.85
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CREBtide is a synthetic substrate for PKA (Km = 3.9 µM). This peptide is based on the phosphorylation sequence in δ-CREB (cAMP response element binding protein).
Sequence:KRREILSRRPSYR
MW:1717 Da
% peak area by HPLC:95
Storage condition:-20° C

Specifications

Specifications

  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Cat. No.:
    103003-190
  • Molecular Weight:
    1717 Da
  • Purity:
    95%
  • Protein Synonyms:
    PKA substrate|Protein kinase A substrate|cAMP response element binding protein substrate|CREB substrate
  • Protein/Peptide Name:
    CREBtide
  • Conjugation:
    Unconjugated
  • Sequence:
    KRREILSRRPSYR

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.