To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Size:3.75 mg
- Storage Conditions:–20 °C
- Protein Synonyms:CMV CEF control Peptide pool
- Protein/Peptide Name:Epstein-Barr Virus (EBV) Control Peptide Pool
- Purity:95%
- Tested Applications:Control Peptide
- Cat. No.:103007-300
- Supplier no.:AS-62341
These 15 Cytomegalovirus (CMV) peptides, at 0.25 mg each (total of 3.75 mg/vial), constitute part of the CEF control peptide pool (cat# 61036-025). Now available separately as EBV Control Peptide Pool, CMV Control Peptide Pool (cat# 62339), Influenza control peptide pool (cat# 62340), these peptides have been used in the stimulation of IFNgamma release from CD8+ T cells in individuals with defined HLA types, these epitopes are useful in applications such as ELISPOT, intracellular cytokine and CTL assays. All peptides are provided as net weight based on peptide content.
Sequence:
MW:
% peak area by HPLC:95
Storage condition:-20° C