Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Cytomegalovirus (CMV)
- Size:1.25 mg
- Storage Conditions:–20 °C
- Protein Synonyms:CMV CEF control Peptide pool
- Protein/Peptide Name:Cytomegalovirus (CMV) Control Peptide Pool
- Purity:95%
- Tested Applications:Control Peptide
- Cat. No.:103007-296
- Supplier no.:AS-62339
These 5 Cytomegalovirus (CMV) peptides, at 0.25 mg each (total of 1.25 mg/vial), constitute part of the CEF control peptide pool (cat# AS-61036-025). Now available separately as CMV control peptide pool, Influenza control peptide pool (cat# AS-62340) and EBV control peptide pool (cat# AS-62341), these peptides have been used in the stimulation of IFNgamma release from CD8+ T cells in individuals with defined HLA types, these epitopes are useful in applications such as ELISPOT, intracellular cytokine and CTL assays. All peptides are provided as net weight based on peptide content.
Sequence:
MW:
% peak area by HPLC:95
Storage condition:-20° C