Order Entry
United States
ContactUsLinkComponent
Bacterial Sortase Substrate III, DNP(2, 4-Dinitrophenol)
Bacterial Sortase Substrate III, DNP(2, 4-Dinitrophenol)
Catalog # 103007-528
Supplier:  Anaspec Inc
CAS Number:  
Bacterial Sortase Substrate III, DNP(2, 4-Dinitrophenol)
Catalog # 103007-528
Supplier:  Anaspec Inc
Supplier Number:  AS-63717
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    DNP(2,4-Dinitrophenol)
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    SrtA Peptide|Staphylococcus aureus transpeptidase Sortase A|Sortase
  • Protein/Peptide Name:
    Bacterial Sortase Substrate III
  • Purity:
    95%
  • Molecular Weight:
    928 Da
  • Sequence:
    Abz-LPETG-K(Dnp)-NH2
  • Cat. No.:
    103007-528
  • Supplier no.:
    AS-63717

Specifications

About this item

The Staphylococcus aureus transpeptidase Sortase A (SrtA) anchors virulence and colonization-associated surface proteins to the cell wall. SrtA selectively recognizes a C-terminal LPXTG motif. SrtA readily reacts with its native substrate Abz-LPETG-Dap(DNP)-NH2, cleaving it and catalyzing the formation of an amide bond between the carboxyl group of threonine and the amino group of cell-wall crossbridges. Cleavage of this FRET substrate can be monitored at Ex/Em=320 nm/420 nm.
Sequence:Abz-LPETG-K(Dnp)-NH2
MW:928 Da
% peak area by HPLC:95
Storage condition:-20° C