About this item
This 14-mer prosaptide sequence is derived from the active neurotrophic region in the amino-terminal portion of the saposin C domain. Synthetic peptides derived from this region are biologically active and are named “prosaptides.” Prosaposin and prosaptides are active on a variety of neuronal cells, stimulating sulfatide synthesis and increasing sulfatide concentration in Schwann cells and oligodendrocytes. This indicates that prosaposin and prosaptides are trophic factors for myelin formation.
Sequence:TaLIDNNATEEILY
MW:1579.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Storage Conditions:–20 °C
- Protein Synonyms:Saposin Peptide|TX14(A) Peptide|Prosaptide Peptide
- Protein/Peptide Name:Prosaptide TX14(A) Peptide
- Purity:95%
- Molecular Weight:1579.7 Da
- Sequence:TaLIDNNATEEILY