Order Entry
United States
ContactUsLinkComponent
Cyclo (-RGDyK)
Cyclo (-RGDyK)
  103006-396
 :  
Cyclo (-RGDyK)
  103006-396
 :  AS-61183-5
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    RGD Peptide|RGDyK
  • Protein/Peptide Name:
    Cyclo (-RGDyK)
  • Purity:
    95%
  • Molecular Weight:
    619.7 Da
  • Sequence:
    Cyclo(-RGDyK)
  • Cat. No.:
    103006-396
  • Supplier no.:
    AS-61183-5

 

 

This cyclic RGD peptide contains a subsituted d-Tyr instead of d-Phe on the fourth position. Substitution with Tyr results in a high affinity and selectivity for the avb3 integrin. Substitution with Tyr also allows for electrophilic radiohalogenation if desired.
Sequence:Cyclo(-RGDyK)
MW:619.7 Da
% peak area by HPLC:95
Storage condition:-20° C