Order Entry
United States
ContactUsLinkComponent
4N1K, AnaSpec
4N1K, AnaSpec
Catalog # 103008-256
Supplier:  Anaspec
CAS Number:  
4N1K, AnaSpec
Catalog # 103008-256
Supplier:  Anaspec
Supplier Number:  AS-64591
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Integrin associated protein Peptide|IAP agonist Peptide|cell-binding domain adhesive Peptide
  • Protein/Peptide Name:
    4N1K
  • Purity:
    95%
  • Molecular Weight:
    1384.8 Da
  • Sequence:
    KRFYVVMWKK
  • Cat. No.:
    103008-256
  • Supplier no.:
    AS-64591

Specifications

About this item

4N1K is a cell-binding domain adhesive peptide. It has been identified as an IAP agonist. Integrin-associated protein (IAP) is important in host defense where it is required for integrin-dependent functions of polymorphonuclear leukocytes. IAP also appears to be important in modulating integrin function in other cells and in signal transduction upon ligand binding by certain integrins with which it associates.
Sequence:KRFYVVMWKK
MW:1384.8 Da
% peak area by HPLC:95
Storage condition:-20° C