Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:FAM (Carboxyfluorescein)
- Size:0.1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:MMP Substrate standard|Matrix Metalloproteinase substrate standard|MMP FRET standard
- Protein/Peptide Name:5-FAM MMP FRET Peptide Fluorescence Standard I
- Purity:95%
- Molecular Weight:586.6 Da
- Sequence:5-FAM-PL
- Cat. No.:103005-948
- Supplier no.:AS-60584-01
Matrix Metalloproteinases (MMPs) are a large family of endopeptidases. Collectively, MMPs can degrade all kinds of extracellular matrix proteins, and can also process a number of bioactive molecules. They are known to be involved in the cleavage of cell surface receptors, the release of apoptotic ligands, and chemokine/cytokine inactivation. MMPs are also thought to play a major role in cell behaviors such as cell proliferation, migration (adhesion/dispersion), differentiation, angiogenesis, apoptosis, and host defense.
This peptide contains 5-FAM only and is similar to the proteolytic product of the 520 MMP FRET substrates. It can be used to set up the fluorescence standard curve. Abs/Em = 494/521 nm.
Sequence:5-FAM-PL
MW:586.6 Da
% peak area by HPLC:95
Storage condition:-20° C