Order Entry
United States
ContactUsLinkComponent
Bovine;Human;Mouse;Rat Orexin A
Bovine;Human;Mouse;Rat Orexin A
Catalog # 103003-738
Supplier:  Anaspec Inc
CAS Number:  
Bovine;Human;Mouse;Rat Orexin A
Catalog # 103003-738
Supplier:  Anaspec Inc
Supplier Number:  AS-24470
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Source:
  • Species:
    Human
    Bovine
    Rat
    Mouse
  • Size:
    1 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Orexzin Peptide|Orexin hypothalamic Peptide
  • Protein/Peptide Name:
    Orexin A
  • Purity:
    95%
  • Molecular Weight:
    3561.2 Da
  • Sequence:
    Pyr-PLPDCCRQKTCSCRLYELLHGAGNHAAGILTL-NH2 (Disulfide bridge: 6-12 and 7-14)
  • Cat. No.:
    103003-738
  • Supplier no.:
    AS-24470

Specifications

About this item

This hypothalamic neuropeptide has been shown to regulate feeding behavior.
Sequence:Pyr-PLPDCCRQKTCSCRLYELLHGAGNHAAGILTL-NH2 (Disulfide bridge: 6-12 and 7-14)
MW:3561.2 Da
% peak area by HPLC:95
Storage condition:-20° C