Order Entry
United States
ContactUsLinkComponent
Human Luteinizing Hormone-Releasing Hormone
Human Luteinizing Hormone-Releasing Hormone
Catalog # 102996-122
Supplier:  Anaspec Inc
CAS Number:  
Human Luteinizing Hormone-Releasing Hormone
Catalog # 102996-122
Supplier:  Anaspec Inc
Supplier Number:  AS-20782
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Source:
  • Species:
    Human
  • Size:
    25 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    FSH-RH Peptide|GnRH Peptide|LHRH Peptide
  • Protein/Peptide Name:
    Luteinizing Hormone-Releasing Hormone
  • Purity:
    95%
  • Molecular Weight:
    1182.3 Da
  • Sequence:
    Pyr-HWSYGLRPG-NH2
  • Cat. No.:
    102996-122
  • Supplier no.:
    AS-20782

Specifications

About this item

Luteinizing Hormone-Releasing Hormone (LHRH), also known as GnRH or FSH-RH, is a hypothalamic hormone that controls the release of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) in the pituitary gland and promotes mating behavior. It stimulates the secretion of gonadotropins after its interaction with specific G-protein-coupled receptors that are present in plasma membranes of pituitary gonadotrophs. In addition to hypophysiotropic effect, it also plays a autocrine/paracrine role in peripheral organs such as prostate, breast and ovary. LHRH is not only a neurohormone but may also function as a neurotransmitter or neuromodulator. It also has a role in angiogenesis, cell proliferation, and tumor metastasis.
Sequence:Pyr-HWSYGLRPG-NH2
MW:1182.3 Da
% peak area by HPLC:95
Storage condition:-20° C