ContactUsLinkComponent
Accessories for TriggerGrip™ Dispenser, MicroCare
Accessories for TriggerGrip™ Dispenser, MicroCare
Catalog # 76020-150
Supplier:  MicroCare
undefined
Accessories for TriggerGrip™ Dispenser, MicroCare
Catalog # 76020-150
Supplier:  MicroCare
Supplier Number:  MCC-BK3
Specifications
  • Description:
    Bench Mounting Kit for TriggerGrip™
  • Cat. No.:
    76020-150
Specifications
About this item
This device holds the both the aerosol can and the TriggerGrip™ dispenser neatly at the workbench; keeps the can from tipping over during use; ensures the aerosol can will empty completely.