ContactUsLinkComponent
Composite Lint-Free Wipe, Medium Size, MicroCare
Composite Lint-Free Wipe, Medium Size, MicroCare
Catalog # 76020-246
Supplier:  MicroCare
undefined
Composite Lint-Free Wipe, Medium Size, MicroCare
Catalog # 76020-246
Supplier:  MicroCare
Supplier Number:  MCC-W88
Specifications
  • Description:
    Composite Lint-Free Wipe, Medium Size
  • Cat. No.:
    76020-246
Specifications
About this item
This wipe offers great cleaning at low cost.
  • Heavy economy wipe
  • Great cleaning at low cost
  • Five layers of material in each wipe
  • Class 100 clean room packaging
Great cleaning at low cost. Heavy wipe: 4.0 oz. with five layers of material in each wipe. Resists damage from abrasion.
It is made of five (5) layers of different materials, which produces a wipe that is very absorbent, soft and fluffy. The wipe is 'apertured' (punched with microscopic holes) to enhance absorbency. The wipe is very strong and resists abrasion, so it’s an excellent choice when hard scrubbing is required.
Packaging: 100 sheets/bag, 12 bags/case.