Order Entry
United States
ContactUsLinkComponent
Pierce 1-Step™ NBT/BCIP Substrate Solution, Thermo Scientific
Pierce 1-Step™ NBT/BCIP Substrate Solution, Thermo Scientific
Catalog # PI34042
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce 1-Step™ NBT/BCIP Substrate Solution, Thermo Scientific
Catalog # PI34042
Supplier:  Invitrogen
Supplier Number:  34042
CAS Number:  

Specifications

  • Description:
    1-Step™ NBT/BCIP Substrate Solution
  • Size:
    250 mL
  • Cat. No.:
    PI34042
  • Application:
    Immunoblotting
  • Supplier no.:
    34042
  • No. of uses:
    25 uses at 10 ml each

Specifications

About this item

The 1-Step™ NBT and BCIP substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.

  • Sensitive with low background and minimal assay-to-assay variability
  • Ready-to-use single component

The combination of NBT and BCIP yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, an enzyme conjugate for antibody probes. Used for immunoblotting, immunohistochemistry, and in situ hybridization applications.