To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Source:
- Species:Chicken
- Size:1 mg
- Environmentally Preferable:
- Storage Conditions:–20 °C
- Protein Synonyms:OVA 323-339 FITC|OVA Peptide FITC
- Protein/Peptide Name:OVA (323-339)
- Purity:95%
- Molecular Weight:2276.5 Da
- Sequence:FITC-LC-ISQAVHAAHAEINEAGR
- Cat. No.:103008-438
- Supplier no.:AS-64766
Specifications
About this item
This fluorescent (FITC)-labeled OVA (323 - 339) Peptide is an H-2b-restricted OVA class II epitope (Abs/Em = 493/522 nm).
Sequence: FITC-LC-ISQAVHAAHAEINEAGR
MW: 2276.5 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C