3M™ Zeta Plus™ BC Series Filter Capsule with ZB Series Media
Supplier: 3M Healthcare
Certificates
About this item
Totally disposable capsule depth filters for laboratory, process development, scale-up or small lot production using ZB media.
- Zeta Plus™ BC units are available in three sizes with industry-leading Zeta Plus™ depth filter media to provide a high degree of application flexibility
- ZB Grade Cellulose Pre-extracted (acid washed) Diatomaceous Earth (D.E.)30ZB, 60ZB,90ZB
- Endotoxin reduction in high purity pharmaceutical water systems
The Zeta Plus™ BC Series is a range of completely encapsulated disposable depth filter assemblies, designed for small volume biological, bioprocess and pharmaceutical Filtration.
Specifications
- Catalog No:
- 76029-561
- 76029-563
- 76029-543
- 76029-645
- 76029-533
- 76029-647
- 76029-635
- 76032-141
- 76032-143
- 76029-571
- Max. Operating Temperature:
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C