ContactUsLinkComponent
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 10 Gal. Drawdown Tank, 5598407
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 10 Gal. Drawdown Tank, 5598407
  76031-644
undefined
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 10 Gal. Drawdown Tank, 5598407
  76031-644
 :  7000050389
  • Cat. No.:
    76031-644
  • Filter type:
    Tank
3M™ commercial reverse osmosis water storage tanks, model 10 gal. Drawdown tank is designed for use in steamer applications.
  • Reverse osmosis 10 gallon drawdown tank at 60 psi with 15 psi pre-charge
  • Includes connection kit
  • Drawdown Tank is a reverse osmosis storage tank that includes a connection kit.